Learn More
Invitrogen™ MRC2 Recombinant Rabbit Monoclonal Antibody (5I0I9)

Rabbit Recombinant Monoclonal Antibody
176.00 € - 436.00 €
Spezifikation
| Antigen | MRC2 |
|---|---|
| Klon | 5I0I9 |
| Konzentration | 1.08 mg/mL |
| Inhalt und Lagerung | -20°C, Avoid Freeze/Thaw Cycles |
| Anwendungen | ELISA, Western Blot |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
30283423
|
Invitrogen™
MA557975 |
20 μL |
176.00 €
20 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
30283411
![]() |
Invitrogen™
MA557976 |
100 μL |
436.00 €
100 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Immunogen sequence: DAALPEPNVF LIFSHGLQGC LEAQGGQVRV TPACNTSLPA QRWKWVSRNR LFNLGTMQCL GTGWPGTNTT ASLGMYECDR EALNLRWHCR TLGDQLSLLL GARTSNISKP GTLERGDQTR SGQWRIYGSE EDLCALPYHE VYTIQGNSHG KPCTIPFKYD NQWFHGCTST GREDGHLWCA TTQDYGKDER WGFCPIKSND CETFWDKDQL TDSCYQFNFQ STLSWREAWA SCEQQGADLL SITEIHEQTY INGLLTGYSS TLWIGLNDLD TSGGWQWSDN SPLKYLNWES DQPDNPSEEN CGVIRTESSG GWQNRDCSIA LPYVCKKKPN ATAEPTPPDR WANVKVECEP SWQPFQGHCY RLQAEKRSWQ ESKKACLRGG GDLVSIHSMA ELEFITKQIK QEVEELWIGL NDLKLQMNFE WSDGSLVSFT HWHPFEPNNF RDSLEDCVTI WGPEGRWNDS PCNQSLPSIC KKAGQLSQG.
May play a role as endocytotic lectin receptor displaying calcium-dependent lectin activity. Internalizes glycosylated ligands from the extracellular space for release in an endosomal compartment via clathrin-mediated endocytosis. May be involved in plasminogen activation system controlling the extracellular level of PLAUR/PLAU, and thus may regulate protease activity at the cell surface. May contribute to cellular uptake, remodeling and degradation of extracellular collagen matrices. May play a role during cancer progression as well as in other chronic tissue destructive diseases acting on collagen turnover. May participate in remodeling of extracellular matrix cooperating with the matrix metalloproteinases (MMPs).Spezifikation
| MRC2 | |
| 1.08 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q9UBG0 | |
| 9902 | |
| Recombinant Protein (or fragment). | |
| Antibody |
| 5I0I9 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| MRC2 | |
| CD280; CLEC13E; collagen-binding factor Endo180; C-type lectin domain family 13 member E; C-type mannose receptor 2; Endo180; endocytic receptor (macrophage mannose receptor family); endocytic receptor 180; KIAA0709; Lectin lambda; macrophage mannose receptor 2; mannose receptor C type 2; mannose receptor, C type 2; mKIAA0709; Mrc2; novel lectin; UPARAP; UPAR-associated protein; urokinase-type plasminogen activator receptor-associated protein | |
| MRC2 | |
| Primary | |
| Protein A |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.