Learn More
MSH2 Antibody (CL13724) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-24573B
Product Code. 30299235
  • 737.00 € / 0.10 Milliliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
This item is not returnable. View return policy
Description
Description
MSH2 Monoclonal antibody specifically detects MSH2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MSH2
Monoclonal
Biotin
COCA1, DNA mismatch repair protein Msh2, FCC1, hMSH2, HNPCC, HNPCC1mutS (E. coli) homolog 2 (colon cancer, nonpolyposis type 1), LCFS2, mutS homolog 2, colon cancer, nonpolyposis type 1 (E. coli), MutS protein homolog 2
This antibody was developed using a synthetic peptide derived from P43246, with the exact immunogen sequence remaining proprietary.
0.1 mL
Breast Cancer, Cancer, Core ESC Like Genes, DNA Repair, Mismatch Repair, Stem Cell Markers, Tumor Suppressors
4436
Store at 4⪚C in the dark.
IgG2b
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL13724
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications