missing translation for 'onlineSavingsMsg'
Learn More

MSH2 Antibody (MSH2/6549R), Allophycocyanin, Novus Biologicals™

Artikelnummer. 18660757 Shop All Bio Techne Products
Change view
Click to view available options
Menge:
0.1 mL
Unit Size:
0.10 Milliliter
Product Code. Menge unitSize
18660757 0.1 mL 0.10 Milliliter
1 options
This item is not returnable. View return policy

Product Code. 18660757

Brand: Novus Biologicals NBP308958APC

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

MSH2 Monoclonal antibody specifically detects MSH2 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSH2
Anwendungen Western Blot, Immunohistochemistry (Paraffin)
Klassifikation Monoclonal
Klon MSH2/6549R
Konjugat APC
Zusammensetzung PBS
Gen-Alias COCA1, DNA mismatch repair protein Msh2, FCC1, hMSH2, HNPCC, HNPCC1mutS (E. coli) homolog 2 (colon cancer, nonpolyposis type 1), LCFS2, mutS homolog 2, colon cancer, nonpolyposis type 1 (E. coli), MutS protein homolog 2
Wirtsspezies Rabbit
Immunogen Recombinant full-length human MSH2 protein (Uniprot: IDP43246)
Reinigungsverfahren Protein A or G purified
Menge 0.1 mL
Regulatorischer Status RUO
Forschungsgebiet Breast Cancer, Cancer, Core ESC Like Genes, DNA Repair, Mismatch Repair, Stem Cell Markers, Tumor Suppressors
Primär oder sekundär Primary
Gen-ID (Entrez) 4436
Zielspezies Human
Inhalt und Lagerung Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.