Beschreibung
MSH2 Monoclonal antibody specifically detects MSH2 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Spezifikation
Spezifikation
| Antigen | MSH2 |
| Anwendungen | Western Blot, Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Klon | MSH2/6549R |
| Konjugat | FITC |
| Zusammensetzung | PBS |
| Gen-Alias | COCA1, DNA mismatch repair protein Msh2, FCC1, hMSH2, HNPCC, HNPCC1mutS (E. coli) homolog 2 (colon cancer, nonpolyposis type 1), LCFS2, mutS homolog 2, colon cancer, nonpolyposis type 1 (E. coli), MutS protein homolog 2 |
| Wirtsspezies | Rabbit |
| Immunogen | Recombinant full-length human MSH2 protein (Uniprot: IDP43246) |
| Reinigungsverfahren | Protein A or G purified |
| Mehr anzeigen |