Learn More
Invitrogen™ MSP Recombinant Rabbit Monoclonal Antibody (24GB635)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557054
Beschreibung
MSP Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
MSP (or MST1) contains four kringle domains and a serine protease domain, similar to that found in hepatic growth factor. Despite the presence of the serine protease domain, the encoded protein may not have any proteolytic activity. The receptor for this protein is RON tyrosine kinase, which upon activation stimulates ciliary motility of ciliated epithelial lung cells. This protein is secreted and cleaved to form an alpha chain and a beta chain bridged by disulfide bonds.
Spezifikation
| MSP | |
| Recombinant Monoclonal | |
| Unconjugated | |
| MST1 | |
| D3F15S2; D3F15S2h; D8h3f15s2; D9H3F15S2; DNF15S2; DNF15S2h; E2F transcription factor 2; E2F2; Hepatocyte growth factor-like protein; Hepatocyte growth factor-like protein alpha chain; Hepatocyte growth factor-like protein beta chain; hepatocyte growth factor-like protein homolog; Hgfl; macrophage stimulating 1; macrophage stimulating 1 (hepatocyte growth factor-like); macrophage stimulatory protein; Macrophage-stimulating protein; MSP; Mst1; NF15S2; Unknown (protein for MGC:137619) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 15235, 24566, 4485 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB635 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P26927, P26928 | |
| MST1 | |
| A synthesized peptide derived from human MST1 (aa 15-65). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |