missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MSX1 Monoclonal Antibody (801601)
Mouse Monoclonal Antibody
Marke: Invitrogen™ MA561514
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
MSX1 is a member of the muscle segment homeobox gene family. It functions as a transcriptional repressor during embryogenesis through interactions with components of the core transcription complex and other homeoproteins. MSX1 may also have roles in limb-pattern formation, craniofacial development, particularly odontogenesis, and tumor growth inhibition. Mutations in the gene, which was once known as homeobox 7, have been associated with nonsyndromic cleft lip with or without cleft palate 5, Witkop syndrome, Wolf-Hirschom syndrome, and autosomoal dominant hypodontia.
Spezifikation
| MSX1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P28360 | |
| MSX1 | |
| E. coli-derived recombinant human MSX1, Met1-Thr165. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin) | |
| 801601 | |
| Unconjugated | |
| MSX1 | |
| AA675338; AI324650; ECTD3; homeo box, msh-like 1; homeobox 7; homeobox protein Hox-7; Homeobox protein MSX-1; homeobox, msh-like 1; Hox7; Hox-7; Hox7.1; hox-7.1; HYD1; msh; msh homeo box 1; msh homeobox 1; msh homeobox 1-like protein; msh homeobox homolog 1; Msx1; muscle-segment homeobox; STHAG1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 4487 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur