missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTF2 Polyclonal Antibody

Sheep Polyclonal Antibody
Marke: Invitrogen™ PA5149261
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute at 0.2 mg/mL in sterile PBS.
Polycomb group (PcG) that specifically binds histone H3 trimethylated at 'Lys-36' (H3K36me3) and recruits the PRC2 complex. Acts by binding to H3K36me3, a mark for transcriptional activation, and recruiting the PRC2 complex, leading to enhance PRC2 H3K27me3 methylation activity. Regulates the transcriptional networks during embryonic stem cell self-renewal and differentiation. Promotes recruitment of the PRC2 complex to the inactive X chromosome in differentiating XX ES cells and PRC2 recruitment to target genes in undifferentiated ES cells. Required to repress Hox genes by enhancing H3K27me3 methylation of the PRC2 complex. In some conditions may act as an inhibitor of PRC2 activity: able to activate the CDKN2A gene and promote cellular senescence by suppressing the catalytic activity of the PRC2 complex locally. Binds to the metal-regulating-element (MRE) of MT1A gene promoter.
Spezifikation
| MTF2 | |
| Polyclonal | |
| Unconjugated | |
| Mtf2 | |
| 9230112N11Rik; AA537621; C76717; dJ976O13.2; hPCl2; M96; Metal regulatory transcription factor 2; metal response element binding transcription factor 2; metal response element DNA-binding protein M96; metal response element-binding transcription factor 2; metal-response element DNA-binding protein M96; Metal-response element-binding transcription factor 2; mPCl2; MTF2; PCL2; polycomb-like 2; polycomb-like protein 2; putative DNA binding protein; RGD1304727; RP5-976O13.1; TDRD19A; tudor domain containing 19A; zinc-regulated factor 1; ZiRF1 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 22823 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9Y483 | |
| Mtf2 | |
| E. coli-derived recombinant human MTF2, Asn356-Ser554. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur