Spezifikation
| Menge | 0,1 mL |
| Inhalt und Lagerung | Store at -80C. Avoid freeze-thaw cycles. |
| Forschungskategorie | Signal Transduction |
| Ziel | MTMR4 |
| Synonym | EC 3.1.3.48, FYVE-DSP2, KIAA0647zinc finger, FYVE domain containing 11, myotubularin related protein 4, ZFYVE11FYVE domain-containing dual specificity protein phosphatase 2, Zinc finger FYVE domain-containing protein 11 |
| Gensymbol | MTMR4 |