Learn More
MTUS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech 13436-1-AP-150UL
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 16851654
  • 410.00 € / 150 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
The antibody can recognize all isoforms except isoform 5 (85-90 kDa). The antibody tested HomoDimer isoforms (80 kDa/110-120/280 kDa) in Jurkat cell.
The protein belongs to the FKBP-type peptidyl-prolyl cis/trans isomerase family. It is located in endoplasmic reticulum and acts as molecular chaperones.
Spezifikation
Spezifikation
MTUS1
Polyclonal
Unconjugated
MTUS1
AT2 receptor binding protein, ATBP, ATIP, GK1, KIAA1288, MP44, MTSG1, MTUS1
Rabbit
Antigen Affinity Chromatography
RUO
102103, 306487, 57509
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5HZI1, Q6IMY1, Q9ULD2
MTUS1
MTUS1 Fusion Protein Ag4242
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen