Beschreibung
The MUL1 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Spezifikation
Spezifikation
| Zelllinie | The lysate was created in HEK293T cells, using plasmid ID RC2438 and based on accession number NM_24544. The protein contains a C-terminal DDK tag. |
| Zur Verwendung mit (Anwendung) | Western Blot |
| Menge | 0,1 mg |
| Lagerungspuffer | RIPA buffer |
| Reinheit | Protein |
| Testspezifität | Homo sapiens mitochondrial E3 ubiquitin ligase 1 (MUL1), nuclear gene encoding mitochondrial protein, mRNA. |
| Inhalt und Lagerung | -80°C. Avoid freeze-thaw cycles. |
| Protein State | Native |
Nur für Forschungszwecke