Learn More
Invitrogen™ MVK Recombinant Rabbit Monoclonal Antibody (24GB115)
Beschreibung
MVK Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Mevalonate kinase (MVK) is an early enzyme in isoprenoid and sterol synthesis. Mevalonate kinase catalyzes the ATP-dependent phosphorylation of mevalonic acid to form mevalonate 5-phosphate. Deficiency in MVK activity contributes to mevalonic aciduria and hyperimmunoglobulinemia D/periodic fever syndrome (HIDS).
Spezifikation
Spezifikation
| Antigen | MVK |
| Anwendungen | Flow Cytometry, Western Blot, Immunocytochemistry |
| Klassifikation | Recombinant Monoclonal |
| Klon | 24GB115 |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gen | Mvk |
| Gen-Zugriffsnummer | P17256, Q03426 |
| Gen-Alias | 2310010A05Rik; AI256848; AI414037; KIME; LH receptor mRNA-binding protein; LHR mRNA binding protein; Lrbp; Mevalonate kinase; mevalonate kinase 1; MK; Mvk; MVLK; POROK3 |
| Gensymbole | Mvk |
| Mehr anzeigen |