missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ MxA/Mx1 Antibody (MX1/7529) [Alexa Fluor™ 594], Novus Biologicals™

Mouse Monoclonal Antibody

Marke:  Novus Biologicals™ NBP3-26970AF594

Artikelnummer. 30063948

  • ONLINE EXCLUSIVE PRICE
    713.00 €
    674.10 € / 0.10 Milliliter
    Sparen 38.90 €
    5% Rabatt

om dit product te kopen Registreer vandaag om een webaccount aan te maken

This item is not returnable. View return policy

Description

Description

MxA/Mx1 Monoclonal antibody specifically detects MxA/Mx1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MxA/Mx1
Monoclonal
Alexa Fluor 594
human interferon-regulated resistance GTP-binding protein MXA10IFI-78Khomolog of murine (interferon-inducibleprotein p78), myxovirus (influenza virus) resistance 1, interferon-inducible protein p78(mouse)
Recombinant fragment (around aa400-592) of human MxA/Mx1 protein (exact sequence is proprietary)
0.1 mL
Apoptosis
4599
Store at 4°C in the dark.
IgG2 κ
Immunohistochemistry (Paraffin)
MX1/7529
50 mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.