missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myeloperoxidase/MPO Antibody (CL14991) [CoraFluor™ 1], Novus Biologicals™
Alle ansehen Bio Techne ProdukteBeschreibung
Myeloperoxidase/MPO Monoclonal antibody specifically detects Myeloperoxidase/MPO in Human samples. It is validated for Immunohistochemistry,Western Blot
Spezifikation
Spezifikation
| Antigen | Myeloperoxidase/MPO |
| Anwendungen | Immunohistochemistry, Western Blot |
| Klassifikation | Monoclonal |
| Klon | CL14991 |
| Konjugat | CoraFluor 1 |
| Zusammensetzung | PBS |
| Gen-Alias | EC 1.11.1, EC 1.11.1.7, myeloperoxidase |
| Wirtsspezies | Mouse |
| Immunogen | This antibody was developed using a synthetic peptide derived from P05164, with the exact immunogen sequence remaining proprietary. |
| Reinigungsverfahren | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?