Learn More
Myoglobin Antibody (4E2cc), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NB600-1291AF532
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Myoglobin Monoclonal antibody specifically detects Myoglobin in Human samples. It is validated for Immunoassay, ELISA
Spezifikation
| Myoglobin | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoassay, ELISA | |
| 4E2cc | |
| Immunoassay, Sandwich ELISA | |
| MGC13548, myoglobin, PVALB | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Human cardiac myoglobin. | |
| 0.2 mL | |
| Cancer, Cardiovascular Biology | |
| 4151 | |
| Store at 4C in the dark. | |
| IgG1 |