missing translation for 'onlineSavingsMsg'
Learn More

N-(2,6-Dimethylphenyl)-2,2-dichloroacetamide, TRC

Artikelnummer. 30010863
Change view
Click to view available options
Menge:
100 mg
250 mg
500 mg
Packungsgröße:
100 Milligramm
250 Milligramm
500 Milligramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30010863 250 mg 250 Milligramm
30012714 100 mg 100 Milligramm
30009502 500 mg 500 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010863 Supplier Toronto Research Chemicals Supplier No. TRCD434405250MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39084-88-3
Summenformel C10 H11 Cl2 N O
Molekulargewicht (g/mol) 232.11
Synonym 2,2-Dichloro-N-(2,6-dimethylphenyl)acetamide, N-(2,6-Dimethylphenyl)-2,2-dichloroacetamide, NSC 37288
IUPAC-Name 2,2-dichloro-N-(2,6-dimethylphenyl)acetamide
SMILES Cc1cccc(C)c1NC(=O)C(Cl)Cl

Specifications

Chemischer Name oder Material 2,2-Dichloro-N-(2,6-dimethylphenyl)acetamide
Hinweise zur Reinheitsqualität HPLC
Menge 250 mg
InChI-Formel InChI=1S/C10H11Cl2NO/c1-6-4-3-5-7(2)8(6)13-10(14)9(11)12/h3-5,9H,1-2H3,(H,13,14)
Formelmasse 231.0218
Reinheit (%) >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.