Learn More
CCDC91 Antibody, Allophycocyanin, Novus Biologicals™
Artikelnummer. 18611852 Alle ansehen Bio Techne Produkte
Ansicht ändern
Click to view available options
Packungsgröße:
0.10 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. unitSize
18611852 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18611852 Lieferant Novus Biologicals Lieferanten-Nr. NBP298667APC
Please to purchase this item. Need a web account? Register with us today!
Rabbit Polyclonal Antibody
CCDC91 Polyclonal antibody specifically detects CCDC91 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spezifikation
Immunogen Produced in rabbits immunized with E. coli-derived Human CCDC91 fragment.
Reinigungsverfahren Antigen and protein A Affinity-purified