Learn More
N-Nitroso Trandolapril, TRC
Specifications
Specifications
| Chemischer Name oder Material | N-Nitroso Trandolapril |
| CAS | 1026154-51-7 |
| Summenformel | C24H33N3O6 |
| Menge | 50 mg |
| Synonym | (2S,3aR,7aS)-1-(N-((S)-1-Ethoxy-1-oxo-4-phenylbutan-2-yl)-N-nitroso-L-alanyl)octahydro-1H-indole-2-carboxylic Acid ethyl ester |
| InChI-Formel | InChI=1S/C24H33N3O6/c1-3-33-24(31)20(14-13-17-9-5-4-6-10-17)27(25-32)16(2)22(28)26-19-12-8-7-11-18(19)15-21(26)23(29)30/h4-6,9-10,16,18-21H,3,7-8,11-15H2,1-2H3,(H,29,30)/t16-,18+,19-,20-,21-/m0/s1 |
| SMILES | CCOC([C@@H](N([C@H](C(N1[C@@]2([C@@]([H])(C[C@H]1C(O)=O)CCCC2)[H])=O)C)N=O)CCC3=CC=CC=C3)=O |
| IUPAC-Name | (2S,3aR,7aS)-1-(N-((S)-1-Ethoxy-1-oxo-4-phenylbutan-2-yl)-N-nitroso-L-alanyl)octahydro-1H-indol-2-carbonsäure |
| Molekulargewicht (g/mol) | 459.54 |
| Formelmasse | 459.24 |