Learn More
N-Succinimidyl-S-Acetylthioacetat, TRC
Specifications
Specifications
| Chemischer Name oder Material | N-Succinimidyl-S-acetylthioacetate |
| CAS | 76931-93-6 |
| Summenformel | C8 H9 N O5 S |
| Menge | 100 mg |
| InChI-Formel | InChI=1S/C8H9NO5S/c1-5(10)15-4-8(13)14-9-6(11)2-3-7(9)12/h2-4H2,1H3 |
| SMILES | CC(=O)SCC(=O)ON1C(=O)CCC1=O |
| IUPAC-Name | (2,5-Dioxopyrrolidin-1-yl) 2-Acetylsulfanylacetat |
| Molekulargewicht (g/mol) | 231.23 |
| Formelmasse | 231.0201 |