missing translation for 'onlineSavingsMsg'
Learn More

N2-t-Boc-N6-(Biotinamido-6-N-caproylamido)lysin, TRC

Product Code. 30341612
Change view
Click to view available options
Menge:
10 mg
Unit Size:
10 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
30341612 10 mg 10 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30341612 Supplier Toronto Research Chemicals Supplier No. TRCB64250010MG
 Eingeschränkter Artikel
Verfügbar ab: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material N2-t-Boc-N6-(biotinamido-6-N-caproylamido)lysine
CAS 102910-26-9
Summenformel C27 H47 N5 O7 S
Menge 10 mg
InChI-Formel InChI=1S/C27H47N5O7S/c1-27(2,3)39-26(38)30-19(24(35)36)12-8-10-16-29-20(33)13-5-4-9-15-28-21(34)14-7-6-11-18-17-40-23-22(18)31-25(37)32-23/h18-19,22-23H,4-17H2,1-3H3,(H,28,34)(H,29,33)(H,30,38)(H,35,36)(H2,31,32,37)/t18-,19+,22-,23+/m1/s1
SMILES CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)CCCCCNC(=O)CCCC[C@@H]1CS[C@@H]2NC(=O)N[C@H]12)C(=O)O
IUPAC-Name (2S)-6-[6-[5-[(3aS,6S,6aR)-2-oxo-1,3,3a,5,6,6a-hexahydrothieno[2,3-d]imidazol-6-yl]pentanoylamino]hexanoylamino]-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexansäure
Molekulargewicht (g/mol) 585.76
Formelmasse 585.3196
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.