Learn More
Invitrogen™ Namilumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Marke:  Invitrogen™ MA559304
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 30284091
  • 1262.00 € / 1 Milligramm
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Namilumab has been used in trials studying the treatment of Plaque Psoriasis and Rheumatoid Arthritis.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Namilumab
Recombinant Monoclonal
Unconjugated
MT203
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM Histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen