missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NAMPT Biosimilar Recombinant Monoclonal Antibody

Recombinant Monoclonal Antibody
477.00 € - 1204.00 €
Spezifikation
| Antigen | NAMPT Biosimilar |
|---|---|
| Konzentration | 1 mg/mL |
| Inhalt und Lagerung | -20°C, Avoid Freeze/Thaw Cycles |
| Anwendungen | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
| Klassifikation | Recombinant Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
30282427
![]() |
Invitrogen™
MA560105 |
100 μg |
477.00 €
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
30283021
|
Invitrogen™
MA560106 |
1 mg |
1204.00 €
1 Milligramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term Alt-100.
Visfatin, also known as pre-B cell colony-enhancing factor, is an adipocytokine that is abundantly secreted by visceral adipose tissue, with insulin mimetic effects. Visfatin mimics insulin signaling by binding to the insulin receptor and reducing glucose release from liver cells, while stimulating glucose utilization in adipocytes and myocytes. Visfatin is also invoved in enhancement of cell proliferation, biosynthesis of nicotinamide mono-and dinucleotide and glucose homeostasis.Spezifikation
| NAMPT Biosimilar | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Lyophilized | |
| IgG1 | |
| Human | |
| Primary | |
| Protein A |
| 1 mg/mL | |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| Unconjugated | |
| Human | |
| RUO | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Antibody |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts