missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAPRT1 Antibody (CL0366) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP3-43973
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
NAPRT1 Monoclonal antibody specifically detects NAPRT1 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry
Spezifikation
| NAPRT1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| EC 2.4.2.11, FHA-HIT-interacting protein, FHIP, NAPRTase, nicotinate phosphoribosyltransferase, nicotinate phosphoribosyltransferase domain containing 1, Nicotinate phosphoribosyltransferase domain-containing protein 1, nicotinic acid phosphoribosyltransferase, PP3856 | |
| This antibody was generated using a recombinant protein sequence of Q6XQN6, with the exact immunogen sequence remaining proprietary. | |
| 100 μL | |
| Lipid and Metabolism | |
| 93100 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry | |
| CL0366 | |
| Unconjugated | |
| Lyophilized in 5% Trehalose. | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL. | |
| Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur