Learn More
NCK2 Monoclonal Antibody (OTI4H7), TrueMAB™, OriGene
Beschreibung
NCK2 Monoclonal antibody specifically detects NCK2 in Human samples. It is validated for Western Blot
Spezifikation
Spezifikation
| Antigen | NCK2 |
| Anwendungen | Western Blot |
| Klassifikation | Monoclonal |
| Klon | OTI4H7 |
| Konzentration | 1 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gen | GLRX3 |
| Gen-Zugriffsnummer | O76003 |
| Gen-Alias | 4833426I10Rik; cytoplasmic protein NCK2; Grb4; growth factor receptor-bound protein 4; NCK adaptor protein 2; NCK2; Nck-2; NCKbeta; non-catalytic region of tyrosine kinase adaptor protein 2; noncatalytic region of tyrosine kinase, beta; SH2/SH3 adaptor protein NCK-beta |
| Mehr anzeigen |