Learn More
Invitrogen™ NDUFS2 Recombinant Rabbit Monoclonal Antibody (24GB495)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557108
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
NDUFS2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
The protein encoded by this gene is a core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase. Mammalian mitochondrial complex I is composed of at least 43 different subunits, 7 of which are encoded by the mitochondrial genome, and the rest are the products of nuclear genes. The iron-sulfur protein fraction of complex I is made up of 7 subunits, including this gene product. Complex I catalyzes the NADH oxidation with concomitant ubiquinone reduction and proton ejection out of the mitochondria. Mutations in this gene are associated with mitochondrial complex I deficiency. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Spezifikation
| NDUFS2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Ndufs2 | |
| AL033311; CI-49; CI-49kD; comp; complex 1, mitochondrial respiratory chain, 49-KD subunit; complex I 49kDa subunit; complex I-49kD; EMBL:AAH82067.1}; NADH dehydrogenase (ubiquinone) Fe-S protein 2; NADH dehydrogenase (ubiquinone) Fe-S protein 2, 49kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 2, mitochondrial; NADH:ubiquinone oxidoreductase core subunit S2; NADH-coenzyme Q reductase; NADH-ubiquinone oxidoreductase 49 kDa subunit; NADH-ubiquinone oxidoreductase NDUFS2 subunit; Ndufs2; ndufs2 {ECO:0000312; unnamed protein product | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 226646, 289218, 4720 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 24GB495 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75306, Q641Y2, Q91WD5 | |
| Ndufs2 | |
| A synthesized peptide derived from human NDUFS2 (aa 300-434). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |