missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
NECAB1 Monoclonal antibody specifically detects NECAB1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Spezifikation
Spezifikation
| Antigen | NECAB1 |
| Anwendungen | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Klon | CL0575 |
| Konjugat | Biotin |
| Zusammensetzung | PBS |
| Gen-Alias | EF hand calcium binding protein 1, EFCBP1, EF-hand calcium binding protein 1, EF-hand calcium-binding protein 1, neuronal calcium binding protein, Neuronal calcium-binding protein 1, N-terminal EF-hand calcium binding protein 1, N-terminal EF-hand calcium-binding protein 1, STIP-1, synaptotagmin interacting protein 1 |
| Wirtsspezies | Mouse |
| Immunogen | This antibody was developed using a recombinant protein derived from Q8N987, with the exact immunogen sequence remaining proprietary. |
| Reinigungsverfahren | Protein A purified |
| Mehr anzeigen |
Name des Produkts
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?