missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neemöl, TRC
Specifications
Specifications
| Chemischer Name oder Material | Neem Oil |
| CAS | 8002-65-1 |
| Menge | 500 g |
| Synonym | Azadirachta indica oil, Azadirachta indica seed oil, Margosa oil, Margosa seed oil, Melia azadirachta oil, Melia azadirachta seed oil, Neem Indica, Neem seed oil |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?