Learn More
Tocris Bioscience™ Nelarabine
Click to view available options
Menge:
10 mg
50 mg
Packungsgröße:
10 Milligramm
50 Milligramm
Beschreibung
Nelarabine is a nucleoside analog. Metabolized by adenosine deaminase to active metabolite, arabinofuranosylguanine (ara-G). Inhibits DNA synthesis and induces apoptosis. Inhibits growth of human T cell lymphoblastic leukemia cells in vitro and in vivo.
Spezifikation
Spezifikation
| Chemischer Name oder Material | 9-β-D-Arabinofuranosyl-6-methoxy-9H-purin-2-amine |
| CAS | 121032-29-9 |
| Prozentgehaltsbereich | >98% |
| Menge | 50 mg |
| Summenformel | C11H15N5O5 |
| Formelmasse | Observed MW: 297.27 |