Learn More
Invitrogen™ Neon™ NxT Electroporation System 8-Channel Upgrade Package
Beschreibung
The Neon NxT Electroporation System 8-Channel Upgrade Package is available for the Neon NxT Electroporation System with 1-Channel Pipette (Cat. No. NEON1S and NEON1SK). The upgrade package includes the Neon NxT 8-channel pipette, 8-channel pipette station, and 8-channel software activation.
The Neon NxT Electroporation System 8-Channel Pipette enables the electroporation of 96 samples in less than 15-minutes while allowing for full flexibility over each reaction. Due to the 8-channel tube design, each tip is separated in its own chamber and isolated from the other tips, allowing for different electroporation parameters, resuspension buffer, delivery payload, or even cell type. Additionally, the Neon NxT 8-channel pipette utilizes the same Neon NxT tips and buffers as the 1-channel pipette.
Spezifikation
Spezifikation
| Kompatible Spitzen | Neon™ NxT |
| Anzahl Kanäle | 8 |
| Volumen (metrisch) | 10 μL, 100 μL |
| Zur Verwendung mit (Anwendung) | Transfektion |
| Zur Verwendung mit (Geräte) | Neon™ NxT Electroporation System (Cat. No. NEON1S, NEON1SK) |
| Enthält | Neon™ NxT 8-channel pipette, 8-channel pipette station, 8-channel software activation, and quick reference guide |
| Menge | Je 1 |