missing translation for 'onlineSavingsMsg'
Learn More

Neprilysin/CD10 Antibody (MME/8376R), FITC, Novus Biologicals™

Artikelnummer. 18604364 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.1 mL
Packungsgröße:
0.10 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18604364 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18604364 Leverandør Novus Biologicals Leverandørnr. NBP320771F

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Monoclonal Antibody

Neprilysin/CD10 Monoclonal antibody specifically detects Neprilysin/CD10 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen Neprilysin/CD10
Anwendungen Immunohistochemistry (Paraffin)
Klassifikation Monoclonal
Klon MME/8376R
Konjugat FITC
Zusammensetzung PBS
Gen-Alias Atriopeptidase, CALLAmembrane metallo-endopeptidase (neutral endopeptidase, enkephalinase), CD10 antigen, CD10), CD10membrane metallo-endopeptidase variant 1, Common acute lymphocytic leukemia antigen, DKFZp686O16152, EC 3.4.24, EC 3.4.24.11, Enkephalinase, EPN, membrane metallo-endopeptidase, membrane metallo-endopeptidase variant 2, MGC126681, MGC126707, NEPmembrane metallo-endopeptidase (neutral endopeptidase, enkephalinase, CALLA, neprilysin, Neutral endopeptidase, Neutral endopeptidase 24.11, SFE, Skin fibroblast elastase
Wirtsspezies Rabbit
Immunogen Recombinant human Neprilysin/CD10 protein fragment (aa 1-500) (exact sequence is proprietary)
Reinigungsverfahren Protein A or G purified
Menge 0.1 mL
Regulatorischer Status RUO
Forschungsgebiet B Cell Development and Differentiation Markers, Cancer, Cell Biology, Cytokine Research, Immunology, Stem Cell Markers
Primär oder sekundär Primary
Gen-ID (Entrez) 4311
Zielspezies Human
Inhalt und Lagerung Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.