missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
Nestin Polyclonal antibody specifically detects Nestin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown
Specifications
Specifications
| Antigen | Nestin |
| Anwendungen | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Verdünnung | Western Blot 0.04 - 0.4 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000, Knockdown Validated |
| Zusammensetzung | PBS (pH 7.2), 40% Glycerol |
| Gen-Alias | FLJ21841, nestin |
| Wirtsspezies | Rabbit |
| Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: DPEGQSQQVGAPGLQAPQGLPEAIEPLVEDDVAPGGDQASPEVMLGSEPAMGESAAGAEPGPGQGVGGLGDPGHLTREEVMEPPLEEESLEAKRVQGLEGPRKDLEEAGGLGTEFSELP |
| Reinigungsverfahren | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?