missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Neurofascin Monoclonal Antibody (727030)

Mouse Monoclonal Antibody
Marke: Invitrogen™ MA561279
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
Neurofascin encodes an L1 family immunoglobulin cell adhesion molecule with multiple IGcam and fibronectin domains. The protein functions in neurite outgrowth, neurite fasciculation, and organization of the axon initial segment (AIS) and nodes of Ranvier on axons during early development. Both the AIS and nodes of Ranvier contain high densities of voltage-gated Na+ (Nav) channels which are clustered by interactions with cytoskeletal and scaffolding proteins including this protein, gliomedin, ankyrin 3 (ankyrin-G), and betaIV spectrin. This protein links the AIS extracellular matrix to the intracellular cytoskeleton. Neurofascin undergoes extensive alternative splicing, and the full-length nature of some variants has not been determined.
Spezifikation
| Neurofascin | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and no preservative | |
| O94856, P97685, Q810U3 | |
| NFASC | |
| Mouse myeloma cell line NS0-derived recombinant rat Neurofascin, Ile25-Ala1031. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 727030 | |
| Unconjugated | |
| NFASC | |
| AA387016; D430023G06Rik; DKFZp686P2250; KIAA0756; KIAA0756FLJ46866; mKIAA0756; neurofascin; neurofascin homolog; NF; Nfasc; NRCAML | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 116690, 23114, 269116 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur