missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Neuropoietin Polyclonal Antibody

Goat Polyclonal Antibody
Marke: Invitrogen™ PA5148620
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute at 0.2 mg/mL in sterile PBS.
Neuropoietin is a newly identified member of the IL-6 cytokine family. Members of this family, including IL-6, IL-11, oncostatin M, leukemia inhibitory factor (LIF), cardiotrophin-1 (CT-1), cardiotrophin-like cytokine, and CNTF, display a four-helix bundle structure, and signal through gp130-containing receptor complexes. Neuropoietin, which is predominantly expressed in neuroepithelia during embryonic life, acts through a receptor complex formed of a CNTF receptor-alpha component, gp 130, and LIF receptor. Like CNTF, it promotes the survival of embryonic motor neurons, and could increase the proliferation of neural precursor cells in the presence of EGF and FGF-2. Interestingly, the human neuropoietin gene has evolved toward a pseudogene, suggesting that alternative signaling via CNTF is an effective compensatory pathway.
Spezifikation
| Neuropoietin | |
| Polyclonal | |
| Unconjugated | |
| Ctf2 | |
| cardiotrophin 2; cardiotrophin-2; CT-2; Ctf2; CTF2P; Gm494; Neuropoietin; Np | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 244218 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| P83714 | |
| Ctf2 | |
| E. coli-derived recombinant mouse Neuropoietin/NP, Ala23-Ala204. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction