Learn More
NeutraKine™ IFN-beta Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Marke:  Proteintech 69013-1-IG-25UG
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 30236053
  • ONLINE EXCLUSIVE PRICE
    149.97 €
    149.00 € / 25 Mikrogramm
    Sparen 0.97 €
    0.65% Rabatt
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
The NeutraKine™ IFN beta antibody from Proteintech is a mouse monoclonal antibody generated with recombinantprotein of human NeutraKine™ IFN beta. This antibody recognizes human antigen. The NeutraKine™ IFN beta antibody has been validated for the following applications: ELISA, Neutralization analysis.
Spezifikation
Spezifikation
IFN beta
2C6E5
IFB; IFF; IFNB; MGC96956
RecombinantProtein
25 μg
3456
Lyophilized antibodies are stable for 1 year from the date of receipt if stored between (-20°C) and (-80°C). Upon reconstitution we recommend that the solution can be stored at(4°C) for short term or at(-20°C) to (-80°C) for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.
Liquid
ELISA, Neutralization
Unconjugated
Mouse
Protein G purification
Primary
Human
Neu Antibody
IgG1
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen