missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NF-L Monoclonal antibody specifically detects NF-L in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | NF-L |
| Anwendungen | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Klassifikation | Monoclonal |
| Klon | CL4729 |
| Konjugat | Alexa Fluor 647 |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | 68 kDa neurofilament protein, neurofilament protein, light chain, neurofilament subunit NF-L, Neurofilament triplet L protein, neurofilament, light polypeptide, neurofilament-light, NF68FLJ53642, NF-L, NFLlight polypeptide 68kDa |
| Wirtsspezies | Mouse |
| Immunogen | This antibody was developed using a recombinant protein derived from P07196, with the exact immunogen sequence remaining proprietary. |
| Reinigungsverfahren | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?