Learn More
NFATC2 Rabbit anti-Human, Mouse, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech CL488-22023-100UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
NFATC2 Polyclonal antibody apecifically detetects NFATC2 antigen in Human, Mouse Samples. It is validated for Immunofluorescence, Flow Cytometry.Spezifikation
| NFATC2 | |
| Polyclonal | |
| CoraLite 488 | |
| BC136418 | |
| Rabbit | |
| Antigen affinity purification | |
| Primary | |
| Human, Mouse | |
| Liquid |
| Immunofluorescence, Flow Cytometry | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| NFATC2 | |
| NFATC2 fusion protein Ag16849 | |
| 100 μL | |
| 4773 | |
| Store at -20°C. Avoid exposure to light. | |
| IgG |