missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NFIB Recombinant Rabbit Monoclonal Antibody (24GB3600)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557137
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
NFIB Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
NF-1, also designated CTF, consists of a family of CCAAT box binding proteins that stimulate DNA replication and activate transcription. Analysis of human NF-1 messenger RNA has revealed two forms of the NF-1 protein arising from an alternate splicing of a single NF-1 gene. NF-1 binds its consensus DNA element as a homodimer via an amino-terminal DNA binding domain, and activates transcription through a putatively novel, proline-rich, carboxy terminal transactivation domain. The NF-1 protein has been shown to recognize and bind the adenovirus type 2 promoter and activate transcription of herpes simplex virus thymidine kinase genes. The NF-1 consensus element has been found in the upstream promoter region of myriad eukaryotic genes, including that of Ha-Ras, alpha-globin, HSP 70, GRP 78, Histone H1, myelin basic protein and in the Xenopus laevis vitellogenin gene promoter.
Spezifikation
| NFIB | |
| Recombinant Monoclonal | |
| 1.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O00712, P97863 | |
| NFIB | |
| A synthesized peptide derived from human NFIB / NF1B2 (aa 380-420). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB3600 | |
| Unconjugated | |
| NFIB | |
| 6720429L07Rik; CCAAT-box-binding transcription factor; CTF; E030026I10Rik; HMGIC/NFIB; NF1-B; NF-I/B; Nfib; NFI-B; NFIB2; NFIB3; NFI-RED; nuclear factor 1 B-type; nuclear factor 1/B; nuclear factor I B; nuclear factor I/B; olfactory epithelium nuclear factor I-B; TGGCA-binding protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18028, 29227, 4781 | |
| -20°C | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur