missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFkB2/NFkB p100 Antibody (CL8266) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP3-44219
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
NFkB2/NFkB p100 Monoclonal antibody specifically detects NFkB2/NFkB p100 in Human samples. It is validated for Immunohistochemistry (Paraffin),Immunohistochemistry
Spezifikation
| NFkB2/NFkB p100 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| CVID10;H2TF1;LYT10;LYT-10;NF-kB2;Nuclear factor NF-kappa-B p100 subunit;p100;p49/p100;p52 | |
| This antibody was generated using a synthetic peptide of Q00653, with the exact immunogen sequence remaining proprietary. | |
| 100 μL | |
| Apoptosis, B Cell Development and Differentiation Markers, Cancer, Cell Biology, Immunology, Signal Transduction | |
| 4791 | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin), Immunohistochemistry | |
| CL8266 | |
| Unconjugated | |
| Lyophilized in 5% Trehalose. | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL. | |
| Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur