missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NGFR/TNFRSF16/p75NTR Antibody (2F1C2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
226.00 € - 473.00 €
Spezifikation
| Antigen | NGFR/TNFRSF16/p75NTR |
|---|---|
| Klon | 2F1C2 |
| Verdünnung | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 |
| Anwendungen | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18226623
|
Novus Biologicals
NBP1-51590SS |
0.025 mL |
226.00 €
25 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18240864
|
Novus Biologicals
NBP1-51590 |
0.1 mL |
473.00 €
0.10 Milliliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
NGFR/TNFRSF16/p75NTR Monoclonal specifically detects NGFR/TNFRSF16/p75NTR in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Spezifikation
| NGFR/TNFRSF16/p75NTR | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Ascites with 0.03% Sodium Azide | |
| CD271, CD271 antigen, Gp80-LNGFR, member 16, nerve growth factor receptor, NGF receptor, p75 ICD, TNFRSF16nerve growth factor receptor (TNFR superfamily, member 16), tumor necrosis factor receptor superfamily member 16 | |
| NGFR | |
| IgG1 | |
| Unpurified | |
| 45 kDa |
| 2F1C2 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| P08138 | |
| 4804 | |
| Purified recombinant fragment of human p75NTR/NGF Receptor expressed in E. coli. [UniProt# P08138] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts