Learn More
Nickel Electro™ Clifton™ Lift Off Gable Lid
Reduce evaporation, assist in keeping samples in a water bath free from contamination and save energy
Marke:  Nickel Electro™ GL1-8AF
Artikelnummer. 15693454
  • ONLINE EXCLUSIVE PRICE
    129.00 €
    110.33 € / 1 Stück
    Sparen 18.67 €
    14% Rabatt
Nur noch null auf Lager
Verfügbar ab: 03-02-2026
Beschreibung
  • Designed allowing full use of whole width of a Clifton™ water bath with the lowest height of the gable being 45 mm above the bath
  • This feature means test tube racks or flasks can be placed across the whole width of the bath without fouling the lid, maximising the bath working area
  • Designed with the customer in mind, ensuring where condensation accumulates in use there are no drip points which could spoil work in open flasks in your Clifton™ water bath
  • Water evaporation is reduced at elevated temperatures, maintaining water levels in the Clifton™ water bath
  • Insulated handle
  • Allows extended use of the bath, right into the edges of the tank
  • No drip points to contaminate samples, whilst the lid design ensures all condensate returns back into the bath. Insulated handles for safe use