missing translation for 'onlineSavingsMsg'
Learn More

Nitrosamine Mix 1 2000 μg/ml in Methanol, Dr. Ehrenstorfer

Product Code. 30319055
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Menge:
1 mL
missing translation for 'unitSize'
1 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30319055 1 mL 1 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30319055 missing translation for 'mfr' Dr. Ehrenstorfer missing translation for 'supplierNo' DREYA08070100ME

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Entdecken Sie die von Dr. Ehrenstorfer zertifizierten Referenzmaterialien: Erhältlich in verschiedenen Formaten, einschließlich Mehrkomponenten-Gemische für regulatorische Zwecke, um Ihre Lebensmittel- und Umweltanalysen mit rückverfolgbarer, ISO-zertifizierter Qualität zu unterstützen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material N-Nitrosodimethylamine∣N-Nitroso-di-n-propylamine∣N-Nitroso-diphenylamine
CAS 62-75-9, 621-64-7, 86-30-6
Bezeichnung der Analyten oder Bestandteile N-Nitrosodimethylamine∣N-Nitroso-di-n-propylamine∣N-Nitroso-diphenylamine
Versandbedingung Room Temperature
Zertifizierung/Konformität ISO/IEC 17025
Solvent or Matrix Methanol
Industry Type Food Testing
Menge 1 mL
Concentration or Composition (by Analyte or Components) N-Nitrosodimethylamine 2000 μg/mL∣N-Nitroso-di-n-propylamine 2000 μg/mL∣N-Nitroso-diphenylamine 2000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.