missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKIAMRE Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Spezifikation
| Antigen | NKIAMRE |
|---|---|
| Konzentration | 0.4 mg/mL |
| Anwendungen | Immunohistochemistry (Paraffin), Western Blot |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
16892294
|
Proteintech
13888-1-AP-20UL |
20 μL |
145.00 €
20 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
16882294
|
Proteintech
13888-1-AP-150UL |
150 μL |
410.00 €
150 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
The protein encoded by this gene is a member of cyclin-dependent protein kinase (CDK) family. CDK family members are highly similar to the gene products of Saccharomyces cerevisiae cdc28, and Schizosaccharomyces pombe cdc2, and are known to be important regulators of cell cycle progression. This gene was identified as a gene absent in leukemic patients with chromosome 5q deletion. This loss may be an important determit of dysmyelopoiesis. Alternative splicing results in multiple transcript variants encoding different isoforms.Spezifikation
| NKIAMRE | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| Q8BLF2, Q8IVW4, Q9JM01 | |
| 213084, 51265, 60396 | |
| NKIAMRE Fusion Protein Ag4812 | |
| Primary | |
| -20°C | |
| CDKL3 |
| 0.4 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CDKL3, cyclin dependent kinase like 3, NKIAMRE | |
| CDKL3 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts