Learn More
Invitrogen™ Nkx2.2 Recombinant Rabbit Monoclonal Antibody (24GB715)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557121
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Nkx2.2 Recombinant Monoclonal Antibody for Western Blot, Flow
The protein encoded by this gene contains a homeobox domain and may be involved in the morphogenesis of the central nervous system. This gene is found on chromosome 20 near NKX2-4, and these two genes appear to be duplicated on chromosome 14 in the form of TITF1 and NKX2-8. The encoded protein is likely to be a nuclear transcription factor.
Spezifikation
| Nkx2.2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| NKX2-2 | |
| Drosophila NK2 transcription factor related, locus 2; glycoprotein GP330, renal; homeobox protein NK-2 homolog B; Homeobox protein Nkx-2.2; homeobox protein Nkx2-2; NK2 homeobox 2; NK2 transcription factor related locus 2; NK2 transcription factor related, locus 2; NK2 transcription factor-like protein B; NKX2.2; Nkx-2.2; Nkx2\.2; Nkx2-2; NKX2B; tinman | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18088, 4821 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB715 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95096, P42586 | |
| NKX2-2 | |
| A synthesized peptide derived from human NKX2.2 (aa 1-140). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |