missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKX3-2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 22491-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
BAPX1 is the mammalian homolog of the Drosophila bagpipe homeobox gene and is expressed in the splanchnic mesoderm and embryonic skeleton. It is one of the earliest developmental markers for the sclerotome portion of the somite and the gut mesentery. BAPX1 is required for normal skeletal development; homozygous inactivating mutations in the BAPX1 gene result in spodylo-megaepiphyseal-metaphyseal dysplasia (SMMD). It has also been suggested to play a role in the proper development of the mammalian gut and is required for distal stomach development as part of a BARX1-dependent pathway.Spezifikation
| NKX3-2 | |
| Polyclonal | |
| Unconjugated | |
| NKX3-2 | |
| BAPX1, Homeobox protein Nkx 3.2, NK3 homeobox 2, NKX3 2, NKX3.2, NKX3-2, NKX3B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12020, 579 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.36 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P78367, P97503 | |
| NKX3-2 | |
| NKX3-2 Fusion Protein Ag17581 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur