Learn More
Invitrogen™ NPR3 Recombinant Rabbit Monoclonal Antibody (24GB3585)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557097
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
NPR3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes one of three natriuretic peptide receptors. Natriutetic peptides are small peptides which regulate blood volume and pressure, pulmonary hypertension, and cardiac function as well as some metabolic and growth processes. The product of this gene encodes a natriuretic peptide receptor responsible for clearing circulating and extracellular natriuretic peptides through endocytosis of the receptor. Multiple transcript variants encoding different isoforms have been found for this gene.
Spezifikation
| NPR3 | |
| Recombinant Monoclonal | |
| 1.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P17342, P41740, P70180 | |
| NPR3 | |
| A synthesized peptide derived from human Natriuretic Peptide Receptor C (aa 500-541). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB3585 | |
| Unconjugated | |
| NPR3 | |
| A NPR-C; ANP-C; ANP-CR; ANPRC; ANPR-C; atrial natriuretic peptide clearance receptor; Atrial natriuretic peptide clearence receptor 3; atrial natriuretic peptide C-type receptor; atrial natriuretic peptide receptor 3; atrial natriuretic peptide receptor 3-like; atrial natriuretic peptide receptor type C; atrionatriuretic peptide receptor C; C5orf23; EF-2; guanylate cyclase C; GUCY2B; lgj; longjohn; natriuretic peptide receptor 3; natriuretic peptide receptor C/guanylate cyclase C (atrionatriuretic peptide receptor C); Nppc receptor; Npr3; NPRC; NPR-C; stri; strigosus | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18162, 25339, 4883 | |
| -20°C | |
| Liquid |