Beschreibung
Nrf2 Monoclonal antibody specifically detects Nrf2 in Human samples. It is validated for Western Blot
Spezifikation
Spezifikation
| Antigen | Nrf2 |
| Anwendungen | Western Blot |
| Klassifikation | Monoclonal |
| Klon | OTI8A10 |
| Konjugat | Unconjugated |
| Verdünnung | Western Blot 1:2000 |
| Zusammensetzung | Lyophilized powder ( 1X PBS, pH 7.3,) |
| Gen-Alias | HEBP1, NF-E2-related factor 2, NFE2-related factor 2, NRF2 erythroid derived 2, like 2, nuclear factor (erythroid-derived 2)-like 2, nuclear factor erythroid 2-related factor 2, nuclear factor erythroid-derived 2-like 2 |
| Wirtsspezies | Mouse |
| Immunogen | Human recombinant protein fragment corresponding to amino acids 330-605 of human NFE2L2 (NP_006155) produced in E.coli. |
| Mehr anzeigen |