missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NT-proBNP Antibody (13G12cc), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NB120-14712B-200UL
This item is not returnable.
View return policy
Description
NT-proBNP Monoclonal specifically detects NT-proBNP in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA.
Specifications
| BNP | |
| Monoclonal | |
| Biotin | |
| PBS | |
| NPPB | |
| Synthetic peptide: ETSGLQEQRNHLQGK, corresponding to N terminal amino acids 13-27 of Human proBNP. | |
| Cytokine Research, Immunology, Neuroscience, Neurotransmission | |
| proBNP (13G12). | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, ELISA | |
| 13G12 | |
| Western Blot, ELISA | |
| BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide B, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein | |
| Mouse | |
| 200 μL | |
| 4879 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction