Beschreibung
NT-proBNP Monoclonal antibody specifically detects NT-proBNP in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
Spezifikation
Spezifikation
| Antigen | NT-proBNP |
| Anwendungen | Western Blot, ELISA, Sandwich ELISA |
| Klassifikation | Monoclonal |
| Klon | 13G12cc |
| Konjugat | FITC |
| Zusammensetzung | PBS |
| Gen-Alias | BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein |
| Wirtsspezies | Mouse |
| Immunogen | Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with synthetic NT-proBNP, corresponding to a.a.r. 13-27, conjugated with carrier protein. |
| Reinigungsverfahren | Protein A purified |
| Mehr anzeigen |