Learn More
Invitrogen™ Nuclear Matrix Protein p84 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Marke:  Invitrogen™ PA553922
Artikelnummer. 15711861
  • 582.00 € / 100 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
Immunogen sequence: VFCGRIQLFL ARLFPLSEKS GLNLQSQFNL ENVTVFNTNE QESTLGQKHT EDREEGMDVE EGEMGDEEAP TTCSIPIDYN LYRKFWS Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 46%.
HPR1 is part of the TREX (transcription/export) complex, which includes TEX1 (MIM 606929), THO2 (MIM 300395), ALY (MIM 604171), and UAP56 (MIM 142560).
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Nuclear Matrix Protein p84
Polyclonal
Unconjugated
Thoc1
3110002N20Rik; AW107452; HPR1; hTREX84; I79_014746; LRRGT00070; NMP-84; Nuclear matrix protein p84; P84; P84N5; THO complex 1; THO complex subunit 1; THO complex subunit 1-like protein; tho1; Thoc1
Rabbit
Antigen affinity chromatography
RUO
9984
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96FV9
Thoc1
Recombinant protein corresponding to Human Nuclear Matrix Protein p84. Recombinant protein control fragment (Product #RP-91835).