missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NXPH3 Recombinant Mouse Monoclonal Antibody (K94060_8A12)
Description
Immunogen sequence: EDPERDDHEG QPRPRVPRKR GHISPKSRPM ANSTLLGLLA PPGEAWGILG QPPNRPNHSP PPSAKVKKIF GWGDFYSNIK Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%, Pig - 88%, Cynomolgus monkey - 97%.
May be signaling molecules that resemble neuropeptides. Ligand for alpha-neurexins. Target protein is a member of a family of neuropeptide-like glycoproteins.
Specifications
Specifications
| Antigen | NXPH3 |
| Anwendungen | Immunohistochemistry (Paraffin) |
| Klassifikation | Recombinant Monoclonal |
| Klon | K94060_8A12 |
| Konzentration | 0.30 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 0.5% BSA, 50% glycerol and 0.09% sodium azide |
| Gen | NXPH3 |
| Gen-Zugriffsnummer | O95157 |
| Gen-Alias | KIAA1159; neurexophilin 3; neurexophilin-3; NPH3; Nxph3; UNQ687/PRO1327 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?