missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NY-ESO-1 Recombinant Rabbit Monoclonal Antibody (6U0N7)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA558009
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: MQAEGRGTGG STGDADGPGG PGIPDGPGGN AGGPGEAGAT GGRGPRGAGA ARASGPGGGA PRGPHGGAAS GLNGCCRCGA RGPESRLLEF YLAMPFATPM.
NY-ESO-1, a member of the CT (cancer/testis) family, is a tumor specific shared antigen with distinctive immunogenicity. The molecular basis of T cell-mediated antitumor immunity has been elucidated by the identification of a number of tumor antigens recognized by CD8+ T cells. Among these antigens, NY-ESO-1 is of particular interest because both the cytotoxic T lymphocyte and antibody have been shown to react with this antigen. The NY-ESO-1 has been detected in normal testis and in a range of human tumor types. NY-ESO-1 reactivity in testis is restricted germ cells, particularly in the spermatogonia.
Spezifikation
| NY-ESO-1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P78358 | |
| CTAG1A | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human NY-ESO-1 0. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 6U0N7 | |
| Unconjugated | |
| CTAG1A | |
| Autoimmunogenic cancer/testis antigen NY-ESO-1; cancer/testis antigen 1; cancer/testis antigen 1A; cancer/testis antigen 1-A; cancer/testis antigen 6.1; CT6.1; CTAG; CTAG1; CTAG1A; CTAG1B; ESO1; l antigen family member 2; LAGE2; LAGE-2; LAGE2A; LAGE2B; New York Esophageal Squamous Cell Carcinoma 1; NY-ESO-1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 246100 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur